missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Easy pH Electrodes
A durable, polysulfone pH electrode with an integral bulb protector and gel electrolyte to avoid the inconvenience of refilling in field. Temperature sensors sold separately.
£136.00 - £177.00
Specifications
Junction | Ceramic |
---|---|
Material | Polysulfone |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 80°C |
Type | Combination Electrode |
Product Code | Brand | Model | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | |||
10426463
|
METTLER TOLEDO™
51343011 |
InLab™ Easy BNC | BNC | Fixed Cable to sensor |
£177.00
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
10786723
|
METTLER TOLEDO™
51343010 |
InLab™ Easy | S7 | S7 to BNC (30281915) |
£136.00
Each |
In Stock
Log in to see stock. |
|||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Aquarium water
- Soft drinks
- Beer
- General lab
Accessories & Care
- Ceramic junction
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
- Temperature sensors sold separately (NTC 30 kΩ: 51300164, PT1000: 51300165)
Specifications
Ceramic | |
0 to 14 | |
Combination Electrode | |
Low maintenance, field measurement and educational use | |
U Glass | |
<20 sec. | |
32°F to 176°F | |
4.72 in. | |
0.47 in. |
Polysulfone | |
0°C to 80°C | |
Gel | |
Ag/AgCl | |
NTC 30 kilohm | |
3mol/L KCl | |
0°C to 80°C | |
120 mm | |
12 mm |