missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InLab™742-5m Conductivity Probe

Brand:  Mettler Toledo™ 51344128

Code : 59

Additional Details : Weight : 0.48000kg

Product Code. 11957801

  • £700.00 / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

0.105 cm-1
Conductivity Probe
Epoxy, Graphite
Inlab 742
For Many Different Sample Types
120 mm
Rugged Plastic Shaft
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers