missing translation for 'onlineSavingsMsg'
Learn More

Iniparib, MedChemExpress

Product Code. 16475696
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475696 100 mg 100mg
16454918 10 mg 10mg
16451167 50 mg 50mg
16431626 500 mg 500mg
16431026 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16475696 Supplier Medchem Express Supplier No. HY12015
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Iniparib (BSI-201) is an irreversible inhibitor of PARP1, used in the research of triple negative breast cancer.

Iniparib (BSI-201) is an irreversible inhibitor of PARP1, used in the research of triple negative breast cancer.

Specifications

Chemical Name or Material Iniparib
CAS 160003-66-7
Color Bright Yellow
Purity Grade Notes Research
Molecular Formula C7H5IN2O3
Quantity 100 mg
Synonym BSI-201 NSC-746045 IND-71677
Solubility Information DMSO : 100 mg/mL (342.43 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C([N+]([O-])=O)=C1)I)N
Molecular Weight (g/mol) 292.03
Formula Weight 292.03
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.