missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin beta B Antibody (34B10), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26199-100ul
This item is not returnable.
View return policy
Description
Inhibin beta B Monoclonal antibody specifically detects Inhibin beta B in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| Inhibin beta B | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3625 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA | |
| 34B10 | |
| Flow Cytometry 1:50-1:200, ELISA | |
| activin AB beta polypeptide, Activin beta-B chain, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2, MGC157939 | |
| A synthesized peptide derived from Human Inhibin beta B [UniProt P09529] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction