missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin beta A Antibody (CL15357) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43663-25ul
This item is not returnable.
View return policy
Description
Inhibin beta A Monoclonal antibody specifically detects Inhibin beta A in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Inhibin beta A | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Activin beta-A chain, beta A (activin A, activin AB alpha polypeptide), EDF, erythroid differentiation factor, Erythroid differentiation protein, follicle-stimulating hormone-releasing protein, FRP, FSH-releasing protein, inhibin beta A chain, inhibin, beta A, Inhibin, beta-1 | |
| This antibody was generated using a recombinant protein sequence of P08476, with the exact immunogen sequence remaining proprietary. | |
| 25 μL | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL15357 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3624 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction