missing translation for 'onlineSavingsMsg'
Learn More

Inhibin Antibody (130408R) [Allophycocyanin], Novus Biologicals™

Product Code. 30732521 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30732521 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30732521 Supplier Novus Biologicals Supplier No. FAB338A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Inhibin Monoclonal antibody specifically detects Inhibin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Inhibin
Applications ELISA
Classification Monoclonal
Clone 130408R
Conjugate APC
Formulation PBS
Gene Alias INHA
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Inhibin alpha chain, Accession # P05111
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.