Learn More
Inhibin alpha Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27331-1-AP-150UL
Description
The Inhibin alpha antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Inhibin alpha. This antibody recognizes human, mouse, rat antigen. The Inhibin alpha antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Inhibin alpha | |
| 230 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| INHA, Inhibin alpha, Inhibin alpha chain, inhibin, alpha | |
| 150 μL | |
| 3623 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |