missing translation for 'onlineSavingsMsg'
Learn More

Inhibin alpha Mouse anti-Human, Clone: INHA/4265, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-07434-100ug

 View more versions of this product

Product Code. 18641877

  • £377.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Inhibin alpha
Monoclonal
0.2 mg/mL
Immunohistochemistry-Paraffin 1 to 2 μg/mL
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
A synthetic peptide (around aa 73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary)
100 μg
Apoptosis, Cancer
3623
Store at 4°C.
IgG2a
Immunohistochemistry (Paraffin)
INHA/4265
Unconjugated
1 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.