missing translation for 'onlineSavingsMsg'
Learn More

Inhibin alpha Antibody (INHA/6598R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18384906 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384906 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384906 Supplier Novus Biologicals Supplier No. NBP308782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Inhibin alpha
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone INHA/6598R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 1mM PBS
Gene Alias A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Host Species Rabbit
Immunogen Recombinant fragment of human Inhibin alpha protein (around aa 233-362) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3623
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.