missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INHBB Recombinant Rabbit Monoclonal Antibody (23GB6625), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | INHBB |
|---|---|
| Clone | 23GB6625 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089419
![]() |
Invitrogen™
MA553080 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089748
|
Invitrogen™
MA553081 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INHBB Recombinant Monoclonal antibody specifically detects INHBB in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| INHBB | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P09529, P17491, Q04999 | |
| 16324, 25196, 3625 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6625 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| activin AB beta polypeptide; activin betaB; activin beta-B chain; INHBB; Inhibin beta B chain; inhibin beta B subunit; inhibin beta subunit B; inhibin beta-B; inhibin, beta B; Inhibin, beta-2 | |
| A synthesized peptide derived from human Inhibin beta B (350-407AA). | |
| Primary | |
| -20°C | |
| INHBB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title