missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INHBB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550990
This item is not returnable.
View return policy
Description
INHBB Polyclonal Antibody detects endogenous levels of INHBB protein.
The inhibin beta B subunit joins the alpha subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumour-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. Because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone. Furthermore, the beta B subunit forms a homodimer, activin B, and also joins with the beta A subunit to form a heterodimer, activin AB, both of which stimulate FSH secretion.
Specifications
| INHBB | |
| Polyclonal | |
| Unconjugated | |
| INHBB | |
| activin AB beta polypeptide; activin betaB; activin beta-B chain; INHBB; Inhibin beta B chain; inhibin beta B subunit; inhibin beta subunit B; inhibin beta-B; inhibin, beta B; Inhibin, beta-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16324, 25196, 3625 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| P09529, P17491, Q04999 | |
| INHBB | |
| Synthesized peptide derived from the Internal region of human Inhibin Inhibin B-B (aa 75-124). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction