missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ INHA Recombinant Rabbit Monoclonal Antibody (PD01-29)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556564
This item is not returnable.
View return policy
Description
INHA Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.
Specifications
| INHA | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P05111, P17490, Q04997 | |
| INHA | |
| Synthetic peptide within Human Inhibin alpha aa 233-366. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| PD01-29 | |
| Unconjugated | |
| INHA | |
| A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit | |
| Rabbit | |
| Protein A | |
| RUO | |
| 16322, 24504, 3623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction