Learn More
INHA Monoclonal Antibody (4A2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515315
Description
MA5-15315 targets INHA (Inhibin alpha) in indirect ELISA, IF and IHC applications and shows reactivity with Human samples. The MA5-15315 immunogen is purified recombinant fragment of human INHA expressed in E. Coli. MA5-15315 detects INHA (Inhibin alpha) which has a predicted molecular weight of approximately 40kDa.
The inhibin alpha subunit joins either the beta A or beta B subunit to form a pituitary FSH secretion inhibitor. Inhibin has been shown to regulate gonadal stromal cell proliferation negatively and to have tumor-suppressor activity. In addition, serum levels of inhibin have been shown to reflect the size of granulosa-cell tumors and can therefore be used as a marker for primary as well as recurrent disease. However, in prostate cancer, expression of the inhibin alpha-subunit gene was suppressed and was not detectable in poorly differentiated tumor cells. Furthermore, because expression in gonadal and various extragonadal tissues may vary severalfold in a tissue-specific fashion, it is proposed that inhibin may be both a growth/differentiation factor and a hormone.
Specifications
| INHA | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 0.03% sodium azide | |
| P05111 | |
| INHA | |
| Purified recombinant fragment of human INHA expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry, Immunohistochemistry | |
| 4A2 | |
| Unconjugated | |
| INHA | |
| A inhibin subunit; A-inhibin subunit; AW555078; IHA; Inha; inhibin; inhibin alpha; inhibin alpha chain; inhibin alpha subunit | |
| Mouse | |
| Protein G | |
| RUO | |
| 3623 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.