missing translation for 'onlineSavingsMsg'
Learn More

INH1, MedChemExpress

Product Code. 16437108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437108 10 mg 10mg
16423147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437108 Supplier Medchem Express Supplier No. HY16660

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INH1 specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding. INH1 shows promising cancer inhibition activity both in vitro and in vivo.

INH1 specifically disrupts the Hec1/Nek2 interaction via direct Hec1 binding. INH1 shows promising cancer inhibition activity both in vitro and in vivo.

Specifications

Chemical Name or Material INH1
Physical Form Solid
CAS 313553-47-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H16N2OS
For Use With (Application) Cancer-programmed cell death
Synonym IBT13131
Solubility Information DMSO : 100 mg/mL (324.25 mM; Need ultrasonic)
SMILES O=C(NC1=NC(C2=CC=C(C)C=C2C)=CS1)C3=CC=CC=C3
Molecular Weight (g/mol) 308.4
Formula Weight 308.4
Percent Purity 99.62%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.