missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ingenol Mebutate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity.
Specifications
Specifications
| Chemical Name or Material | Ingenol Mebutate |
| CAS | 75567-37-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C25H34O6 |
| Quantity | 10 mg |
| Synonym | Ingenol 3-angelate PEP005 |
| Solubility Information | DMSO : ≥ 100 mg/mL (232.27 mM) |
| SMILES | C/C=C(C)\C(O[C@H]1C(C)=C[C@]23[C@]1(O)[C@H](O)C(CO)=C[C@@](C3=O)([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C)=O |
| Molecular Weight (g/mol) | 430.53 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction