missing translation for 'onlineSavingsMsg'
Learn More

Ingenol Mebutate, MedChemExpress

Product Code. 16448588
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448588 10 mg 10mg
16421048 5 mg 5mg
16402269 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16448588 Supplier Medchem Express Supplier No. HYB0719
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity.

Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity.

Specifications

Chemical Name or Material Ingenol Mebutate
CAS 75567-37-2
Color White
Purity Grade Notes Research
Molecular Formula C25H34O6
Quantity 10 mg
Synonym Ingenol 3-angelate PEP005
Solubility Information DMSO : ≥ 100 mg/mL (232.27 mM)
SMILES C/C=C(C)\C(O[C@H]1C(C)=C[C@]23[C@]1(O)[C@H](O)C(CO)=C[C@@](C3=O)([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C)=O
Molecular Weight (g/mol) 430.53
Formula Weight 430.53
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.