missing translation for 'onlineSavingsMsg'
Learn More

Ingenol Mebutate, MedChemExpress

Product Code. 16421048
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16421048

Brand: Medchem Express HYB0719

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity.

Ingenol Mebutate is an active ingredient in Euphorbia peplus, acts as a potent PKC modulator, with Kis of 0.3, 0.105, 0.162, 0.376, and 0.171 nM for PKC-α, PKC-β, PKC-γ, PKC-δ, and PKC-ε, respectively, and has antiinflammatory and antitumor activity.

Specifications

Chemical Name or Material Ingenol Mebutate
CAS 75567-37-2
Color White
Purity Grade Notes Research
Molecular Formula C25H34O6
Quantity 5 mg
Synonym Ingenol 3-angelate PEP005
Solubility Information DMSO : ≥ 100 mg/mL (232.27 mM)
SMILES C/C=C(C)\C(O[C@H]1C(C)=C[C@]23[C@]1(O)[C@H](O)C(CO)=C[C@@](C3=O)([H])[C@@]4([H])[C@@](C4(C)C)([H])C[C@H]2C)=O
Molecular Weight (g/mol) 430.53
Formula Weight 430.53
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.