missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10665-1-AP-20UL
This item is not returnable.
View return policy
Description
ING5 is similar to ING1, a tumor suppressor protein that can interact with TP53, inhibit cell growth, and induce apoptosis. This protein contains a PHD-finger, which is a common motif in proteins involved in chromatin remodeling. This protein can bind TP53 and EP300/p300, a component of the histone acetyl transferase complex, suggesting its involvement in TP53-dependent regulatory pathway.Specifications
| ING5 | |
| Polyclonal | |
| Unconjugated | |
| ING5 | |
| ING5, Inhibitor of growth protein 5, p28ING5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 363292, 66262, 84289 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8WYH8, Q9D8Y8 | |
| Ing5 | |
| ING5 Fusion Protein Ag1059 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction