missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Goat anti-Human, Polyclonal, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1681-100UG
This item is not returnable.
View return policy
Description
ING5 Polyclonal specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ING5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA 1:4000 - 1:16000 | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| Synthetic peptide: CGLKKGRGQKEKRGS, corresponding to amino acids 127 - 140 of Human ING5. | |
| 100 μg | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction