missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06217-50ul
This item is not returnable.
View return policy
Description
ING5 Polyclonal antibody specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ING5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000 | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| Produced in rabbits immunized with purified, recombinant Human ING5 (Uniprot#: Q8WYH8-2; Glu36-Gln84) | |
| 50 μg | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction