missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ING5 Polyclonal specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | ING5 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human ING5 (Q8WYH8-2; Glu36-Gln84). |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?