missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING5 Antibody (119), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90261V
This item is not returnable.
View return policy
Description
ING5 Monoclonal specifically detects ING5 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| ING5 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 119 | |
| Western Blot, ELISA | |
| inhibitor of growth family, member 5, inhibitor of growth protein 5, p28ING5FLJ23842 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ING5 (Q8WYH8-2; Glu36-Gln84). | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 84289 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction