missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING4 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67754-1-IG-150UL
This item is not returnable.
View return policy
Description
The ING4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ING4. This antibody recognizes human, pig, rabbit, rat antigen. The ING4 antibody has been validated for the following applications: WB, IHC, IF-P, ELISA analysis.Specifications
| ING4 | |
| Monoclonal | |
| 1000 μg/mL | |
| MGC12557; my036; p29ING4 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Pig, Rabbit, Rat | |
| 29 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1A12A3 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 51147 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction