missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING4, Mouse anti-Human, Clone: 8F7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00051147-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MAAGMYLEHYLDSIENLPFELQRNFQLMRDLDQRTEDLKAEIDKLATEYMSSARSLSSEEKLALLKQIQEAYGKCKEFGDDKVQLAMQTYEMVDKHIRRLDTDLARFEADLKEKQIESSDYDSSSSKGKKKGRTQKEKKAARARSKGKNSDEEAPKTAQKKLKLVRTSPEYGMPSVTFGSVHPSDVLDMPVDPNEPTYCLCHQVSYGEMIGCDNPDCSIEWFHFACVGLTTKPRGKWFCPRCSQERKKKSpecifications
| ING4 | |
| Monoclonal | |
| Unconjugated | |
| inhibitor of growth family, member 4 | |
| MGC12557/my036/p29ING4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 8F7 | |
| In 1x PBS, pH 7.4 | |
| BC007781.2 | |
| ING4 | |
| ING4 (AAH07781.1, 1 a.a. ∼ 249 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51147 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction