missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550229
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ING2 protein.
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.
Specifications
| ING2 | |
| Polyclonal | |
| Unconjugated | |
| Ing2 | |
| 2810011M06Rik; ING1L; ING1Lp; ING2; Ing2b; Inhibitor of growth 1-like protein; inhibitor of growth family member 2; inhibitor of growth family, member 2; inhibitor of growth protein 2; p32; p33ING2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3622, 69260 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9ESK4, Q9H160 | |
| Ing2 | |
| Full length fusion protein of Human ING2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction