missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING2 Monoclonal Antibody (OTI3B11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526476
This item is not returnable.
View return policy
Description
ING2 Monoclonal Antibody for Western Blot
This gene is a member of the inhibitor of growth (ING) family. Members of the ING family associate with and modulate the activity of histone acetyltransferase (HAT) and histone deacetylase (HDAC) complexes and function in DNA repair and apoptosis.
Specifications
| ING2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H160 | |
| Ing2 | |
| Full length human recombinant protein of ING2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI3B11 | |
| Unconjugated | |
| Ing2 | |
| 2810011M06Rik; ING1L; ING1Lp; ING2; Ing2b; Inhibitor of growth 1-like protein; inhibitor of growth family member 2; inhibitor of growth family, member 2; inhibitor of growth protein 2; p32; p33ING2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3622 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction