missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17921-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.Specifications
| ING1 | |
| Polyclonal | |
| Unconjugated | |
| ING1 | |
| ING1, Inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26356, 306626, 3621 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9QXV3, Q9UK53 | |
| Ing1 | |
| ING1 Fusion Protein Ag12338 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction