missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ING1 Monoclonal Antibody (K06325_6F4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556060
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: AAAQADKPNS KRSRRQRNNE NRENASSNHD HDDGASGTPK EKKAKTSKKK KRSKAKAERE ASPADLPIDP NEPTYCLCNQ VSYGEMIGCD NDECPIEWFH FSCVGLNHKP KGKWYCPKCR GENEKTMDKA
This gene encodes a tumor suppressor protein that can induce cell growth arrest and apoptosis. The encoded protein is a nuclear protein that physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Reduced expression and rearrangement of this gene have been detected in various cancers. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
| ING1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9QXV3, Q9UK53 | |
| ING1 | |
| Recombinant human ING1 (immunogen range is 281-410). | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K06325_6F4 | |
| Unconjugated | |
| ING1 | |
| 2610028J21Rik; AA407184; AI875420; growth inhibitor ING1; growth inhibitory protein ING1; ING1; inhibitor of growth family member 1; inhibitor of growth family, member 1; inhibitor of growth protein 1; mING1h; p24ING1c; p33; p33ING; p33ING1; p33ING1b; p33ING1c; p37Ing1b; p47; p47ING1a; tumor suppressor ING1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 26356, 3621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction