missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ING1 Antibody (S03-6B7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19880-100ul
This item is not returnable.
View return policy
Description
ING1 Monoclonal antibody specifically detects ING1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ING1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S03-6B7 | |
| Unconjugated | |
| growth inhibitor ING1, growth inhibitory protein ING1, inhibitor of growth family, member 1, inhibitor of growth protein 1, p24ING1c, p33, p33ING1, p33ING1b, p47, p47ING1a, tumor suppressor ING1 | |
| A synthetic peptide of human ING1 (Uniprot # Q9UK53) | |
| 100 μg | |
| Apoptosis, Cancer, Chromatin Research, p53 Pathway, Transcription Factors and Regulators, Tumor Suppressors | |
| 3621 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction