missing translation for 'onlineSavingsMsg'
Learn More

TPP Techno Plastic Products AG Infors Multitron Adapter Tray 16

Product Code. 17275426
Click to view available options
Holds:
600 mL Bioreactor
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17275426

Brand: TPP Techno Plastic Products AG 87633

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adapter tray for Infors Multitron shaker, made to fit 16 x TubeSpin™ Bioreactors 600 mL

  • TPP adapter tray fit the relevant shaker model from Kuehner or Infors
  • Made of aluminum
  • Guarantee largest possible volume per adapter tray filled with TPP Bioreactor
  • Secure fixation of the tubes with holding bars

Specifications

Holds 600 mL Bioreactor
Tube Capacity 600 mL
Product Type Reusable Tray
Dimensions (L x W x H) 480 x 420 x 139 cm
For Use With (Equipment) Infors Minitron Shaker
No. of Positions 16
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.