missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza B NP Recombinant Rabbit Monoclonal Antibody (004)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£207.00 - £473.00

Specifications

Antigen Influenza B NP
Clone 004
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30535807
GREENER_CHOICE
View Documents
Invitrogen™
MA560369
100 μg
£473.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30534103
View Documents
Invitrogen™
MA560370
20 μg
£207.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Widely recognize Influenza B Nucleoprotein. Has cross-reactivity in ELISA and WB with: Influenza B (B/Austria/1359417/2021) NP Protein; Influenza B (B/Brisbane/60/2008) NP; Influenza B (B/Colorado/06/2017) NP; Influenza B (B/Florida/4/2006) NP; Influenza B (B/Phuket/3073/2013) NP; Influenza B (B/Washington/02/2019) NP. Has cross-reactivity in ELISA with: Influenza B (B/Victoria/705/2018) NP; No cross-reactivity in ELISA and WB with: H1N1 (A/Brevig Mission/1/1918) NP; H1N1 (A/Brisbane/02/2018 NP; H1N1 (A/California/07/2009) NP; H1N1 (A/Guangdong-Maonan/SWL1536/2019) NP; H1N1 (A/Hawaii/70/2019) NP; H1N1 (A/Michigan/45/2015 NP; H1N1 (A/Puerto Rico/8/34/Mount Sinai) NP (I116M); H1N1 (A/Victoria/2570/2019)/(A/Wisconsin/588/2019) NP; H2N2 (A/Ann Arbor/6/1960) NP; H3N2 (A/Aichi/2/1968) NP; H3N2 (A/Cambodia/e0826360/2020 (H3N2)-like NP; H3N2 (A/Hong Kong/1/1968) NP; H3N2 (A/Hong Kong/2671/2019) NP; H3N2 (A/Hong Kong/45/2019) NP; H3N2 (A/Hong Kong/4801/2014) NP; H3N2 (A/Kansas/14/2017) NP; H3N2 (A/Switzerland/9715293/2013) NP; H5N1 (A/Texas/37/2024) NP Protein; H7N9 (A/Anhui/1-BALF_RG6/2013) NP; H7N9 (A/Shanghai/2/2013) NP; No cross-reactivity in ELISA with: H1N1 (A/Victoria/2454/2019) NP; SARS-CoV-2 Nucleocapsid Protein; HCoV-229E Nucleocapsid Protein HCoV-NL63 Nucleocapsid Protein; HCoV-HKU1 Nucleocapsid Protein; HCoV-OC43 Nucleocapsid Protein; No cross-reactivity in WB with: H3N2 (A/Darwin/9/2021)/(A/Darwin/6/2021) NP Protein.

Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
Specifications

Specifications

Influenza B NP
1 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with no preservative
Recombinant Influenza B (B/Florida/4/2006) Nucleoprotein/NP protein (Met1-Tyr560).
Antibody
004
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
Influenza B; Influenza NP
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.