missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza B NP (B/Florida/4/2006) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581758
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza B (B/Florida/4/2006) Nucleoprotein/NP.
Influenza A and B are the two types of influenza viruses that cause epidemic human disease. Influenza B viruses currently circulating are divided into two antigenically and genetically distinct groups: the Victoria and Yamagata lineages. Influenza virus B is a genus in the virus family Orthomyxoviridae. A limited host range means that Influenza virus B pandemics are rare. The nucleoprotein protects the negative strand viral RNA from nucleases by encapsidating it. Encapsidated genomic RNA is termed the ribonucleoprotein (RNP) and serves as template for transcription and replication.
Specifications
| Influenza B NP (B/Florida/4/2006) | |
| Polyclonal | |
| Unconjugated | |
| Influenza B; Influenza NP | |
| Recombinant Influenza B (B/Florida/4/2006) Nucleoprotein/NP Protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction