missing translation for 'onlineSavingsMsg'
Learn More

Influenza B Hemagglutinin Antibody, Alexa Fluor™ 700 - (B/Florida/4/2006), Novus Biologicals™

Product Code. 18333974 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333974 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333974 Supplier Novus Biologicals Supplier No. NBP306576AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Influenza B Hemagglutinin Polyclonal antibody specifically detects Influenza B Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza B Hemagglutinin
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias B1AG46
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Influenza B virus (B/Florida/4/2006) Hemagglutinin (ACA33493.1; Met1-Ala555).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.