missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza B Antibody (1131 (2B2)), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65034H
This item is not returnable.
View return policy
Description
Influenza B Hemagglutinin 2 Monoclonal antibody specifically detects Influenza B Hemagglutinin 2 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications
| Influenza B Hemagglutinin 2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Influenza B virus preparation | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 1131 (2B2) | |
| Immunohistochemistry, Immunohistochemistry-Frozen | |
| Mouse | |
| Protein A purified | |
| RUO | |
| NB100-65034 exhibits nuclear staining in immunofluoresence and is therefore thought to recognize a nuclear protein. Influenza virus type B is a genus in the virus family Orthomyxoviridae responsible for acute respiratory illnesses in humans. It varies antigenically from influenza virus types A and C and is not divided into any subtypes. The gradual evolution of Influenza virus type B occurs via a series of mutations known as antigenic drift, in the surface glycoprotein haemagglutinin (HA), enabling the virus to evade the immune system and potentially re-infect the same host. HA glycoproteins from the Influenza virus type B are included in the annually updated trivalent flu vaccine, along with the Influenza virus type A subtypes H3N2 and H1N1. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction