missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A virus Mouse anti-Influenza virus, Clone: J320, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB13601.1mg
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against influenza A virus.
- High specificity
- High reactivity
Specifications
| Influenza A virus | |
| Monoclonal | |
| Unconjugated | |
| ELISA Immunofluorescence Lateral-Flow Immunochromatographic Assay The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 1 mg | |
| Primary | |
| Influenza virus | |
| Liquid |
| ELISA, Immunofluorescence, Lateral Flow | |
| J320 | |
| Mouse monoclonal antibody raised against influenza A virus. | |
| In 10 mM PBS, pH 7.2 (0.09% sodium azide). | |
| Protein A purification | |
| RUO | |
| This antibody reacts with Nucleoprotein. Reactive with H1 through H14 isolates. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction