Learn More
Influenza A Virus H1N1 Hemagglutinin Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB11396-SP
 View more versions of this product
Product Code. 18746115
  • £200.00 / 25µg
Description
Hemagglutinin/HA Peptide Monoclonal antibody specifically detects Hemagglutinin/HA Peptide in Influenza virusN1 samples. It is validated for Western Blot, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Hemagglutinin/HA Peptide
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
IgG
Western Blot, Immunocytochemistry
2908B
1 μg/mL (Western Blot), 3 to 15 μg/mL (Immunocytochemistry)
YP_009118626
Human embryonic kidney cell, HEK293-derived influenza a virus H1N1 Hemagglutinin with a C-terminal 6-His tag, Met1-Gln529, Accession # YP_009118626
25 μg
Primary
Influenza virus N1
Purified