missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A HA Chimeric Recombinant Rabbit Monoclonal Antibody (CR6261)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551686
This item is not returnable.
View return policy
Description
This antibody binds hemaglutinin of most group 1 influenza viruses. CR6261 recognizes a highly conserved helical region in the membrane-proximal stem of HA1/HA2.
Infectious diseases are the leading cause of death worldwide. AIDS, tuberculosis (TB), malaria, diarrhoeal and respiratory infections account for 78% of deaths caused by infectious disease. As many infectious diseases are controlled, new diseases emerge and old diseases become resistant to current drugs. Many infectious diseases have been associated with an increase risk of carcinoma. Influenza continues to attract researchers as new strains appear by the ability of the influenza gene to mix with different forms of the virus. Recently, research on SARS and West Nile virus has risen due to the increased number of infections. These antibodies assist in research by detecting the infectious disease agent.
Specifications
| Influenza A HA Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG κ |
| Functional Assay, Neutralization | |
| CR6261 | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| The original antibody was isolated using phage display from the IgM+ memory B cells of a healthy individual vaccinated against influenza. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction