missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H9N2 Hemagglutinin anti-Virus, HRP, Antibody Pair, R&D Systems™

Product Code. 18399681
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18399681 15 Plates Pack of 15
18355362 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18399681 Supplier Novus Biologicals Supplier No. NBP30670515PLATES
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Antibody pair for Solid phase sandwich ELISA

Influenza A H9N2 Hemagglutinin specifically detects Influenza A H9N2 Hemagglutinin in Virus samples. It is validated for ELISA/Sandwich ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H9N2 Hemagglutinin
Applications ELISA, Sandwich ELISA
Conjugate HRP
Gene Alias HA, Hemagglutinin, Hemagglutinin HA1 chain, Hemagglutinin HA2 chain
Quantity 15 Plates
Regulatory Status RUO
Target Species Virus
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Type Antibody Pairs
Product Line Novus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.