missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H7N9 NP Recombinant Rabbit Monoclonal Antibody (705)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£132.00 - £357.00

Specifications

Antigen Influenza A H7N9 NP
Clone 705
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30535288
GREENER_CHOICE
View Documents
Invitrogen™
MA560324
100 μL
£357.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30535244
View Documents
Invitrogen™
MA560325
20 μL
£132.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Has cross-reactivity in ELISA with: H7N9 (A/Anhui/1/2013) NP; H7N9 (A/Shanghai/1/2013) NP.

Specifications

Specifications

Influenza A H7N9 NP
1 mg/mL
ELISA
Unconjugated
Rabbit
RUO
PBS with no preservative
Primary
Protein A
705
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
Recombinant Influenza A H7N9 NP protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.