missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H7N9 HA Recombinant Rabbit Monoclonal Antibody (002), Biotin
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£174.00 - £473.00

Specifications

Antigen Influenza A H7N9 HA
Clone 002
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30533736
GREENER_CHOICE
View Documents
Invitrogen™
MA560310
100 μg
£473.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30533748
View Documents
Invitrogen™
MA560311
20 μg
£174.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza A H7N9 HA Recombinant Monoclonal Antibody for ELISA

Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.
Specifications

Specifications

Influenza A H7N9 HA
1 mg/mL
ELISA
Biotin
Rabbit
RUO
PBS with no preservative
Recombinant Influenza A H7N9 HA protein.
Antibody
002
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
flu hemagglutinin; influenza hemagglutinin
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.