Learn More
Invitrogen™ Influenza A H7N9 HA Recombinant Rabbit Monoclonal Antibody (001)

Rabbit Recombinant Monoclonal Antibody
£132.00 - £357.00
Specifications
| Antigen | Influenza A H7N9 HA |
|---|---|
| Clone | 001 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30533749
![]() |
Invitrogen™
MA560312 |
100 μL |
£357.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535102
|
Invitrogen™
MA560313 |
20 μL |
£132.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Has cross-reactivity in ELISA with: H7N9 (A/Anhui/1/2013) HA Protein; H7N9 (A/Hangzhou/1/2013) HA Protein; H7N9 (A/Hangzhou/3/2013) HA Protein; H7N9 (A/Pigeon/Shanghai/S1069/2013) HA Protein; H7N9 (A/Shanghai/1/2013) HA Protein; H7N9 (A/Shanghai/2/2013) HA Protein; H7N9 (A/Shanghai/4664T/2013) HA Protein; H7N9 (A/Zhejiang/1/2013) HA Protein; H7N9 (A/Zhejiang/DTID-ZJU10/2013) HA Protein; H7N7 (A/chicken/Netherlands/1/03) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N2 (A/ruddy turnstone/New Jersey/563/2006) HA Protein; H7N3 (A/turkey/Italy/214845/2002) HA Protein; H7N8 (A/mallard/Netherlands/33/2006) HA Protein; H10N3 (A/duck/Hong Kong/786/1979) HA Protein; H15N8 (A/duck/AUS/341/1983) HA Protein; No cross-reactivity in ELISA with: H1N1 (A/California/07/2009) HA; H1N1 (A/California/04/2009) HA0 Protein; H2N2 (A/Canada/720/2005) HA Protein; H3N2 (A/Brisbane/10/2007) HA Protein; H4N6 (A/mallard/Ohio/657/2002) HA Protein; H5N1 (A/Anhui/1/2005) HA Protein; H6N1 (A/northern shoveler/California/HKWF115/2007) HA Protein; H7N1 (A/turkey/Italy/4602/99) HA1 Protein; H7N3 (A/chicken/SK/HR-00011/2007) HA1 Protein; H7N7 (A/equine/Kentucky/1a/1975) HA Protein; H7N7 (A/Netherlands/219/2003) HA Protein; H7N7 (A/Netherlands/219/2003) HA1 Protein; H8N4 (A/pintail duck/Alberta/114/1979) HA Protein; H9N2 (A/Hong Kong/1073/99) HA Protein; H11N2 (A/duck/Yangzhou/906/2002) HA Protein; H12N5 (A/green-winged teal/ALB/199/1991) HA Protein; H13N8 (A/black-headed gull/Netherlands/1/00) HA Pr...
Influenza A H7N9 HA refers to the hemagglutinin (HA) protein found on the surface of the Influenza A H7N9 virus. Hemagglutinin is one of the major glycoproteins on the influenza virus surface and plays a crucial role in the virus's ability to infect host cells. It facilitates the binding of the virus to the host cell receptors and subsequent entry into the cell. The 'H7N9' designation indicates the specific subtype of the influenza virus, where 'H7' refers to the type of hemagglutinin and 'N9' refers to the type of neuraminidase, another surface protein involved in the release of new virus particles from infected cells. Influenza A H7N9 first emerged in humans in China in 2013 and has been associated with severe respiratory illness and high mortality rates. The HA protein is a primary target for the immune response and is critical for the development of vaccines and antiviral drugs.Specifications
| Influenza A H7N9 HA | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Recombinant Influenza A H7N9 HA protein. | |
| Antibody |
| 001 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| flu hemagglutinin; influenza hemagglutinin | |
| Primary | |
| Protein A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.