missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Influenza A H5N2 HA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581684
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for H5N2 (A/American green-winge teal/California/HKWF609/07) HA/Hemagglutinin.
Specifications
| Influenza A H5N2 HA | |
| Polyclonal | |
| Unconjugated | |
| flu hemagglutinin; influenza hemagglutinin | |
| Recombinant H5N2 (A/American green-winge teal/California/HKWF609/07) HA/Hemagglutinin protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction