missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H5N1 Hemagglutinin Rabbit anti-Influenza virus, Polyclonal, Novus Biologicals™

Product Code. 18352096
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352096 50 μg 50µL
18317023 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18352096 Supplier Novus Biologicals Supplier No. NBP31279650UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Influenza A H5N1 Hemagglutinin Polyclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2μm filtered solution in PBS without Preservative
Host Species Rabbit
Immunogen Purified, recombinant Influenza A H5N1 Hemagglutinin (ABD28180.1; Met1-Gln527)
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Influenza virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.