missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H5N1 Hemagglutinin Mouse anti-Virus, Janelia Fluor 549, Clone: 1E7D8, Novus Biologicals™

Product Code. 18422218 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422218 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422218 Supplier Novus Biologicals Supplier No. NBP175494JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H5N1 Hemagglutinin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E7D8
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.