missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Mouse anti-Virus, DyLight 488, Clone: 1E7D8, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Influenza A H5N1 Hemagglutinin |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 1E7D8 |
| Conjugate | DyLight 488 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | A peptide corresponding to 13 amino acids in the middle of the Hemagglutinin protein. |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?