missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (4E11E1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75505-0.025mg
This item is not returnable.
View return policy
Description
Influenza A Hemagglutinin Monoclonal specifically detects Influenza A Hemagglutinin in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| Influenza A Hemagglutinin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 2.5 μg/mL, ELISA 2 μg/mL | |
| HA epitope tag, HA1, HA2, Hemagglutinin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Virus | |
| IgG1 |
| Western Blot | |
| 4E11E1 | |
| Unconjugated | |
| AAT76166 | |
| HA | |
| Recombinant protein corresponding to amino acids 17 - 338 of H5 HA1. | |
| 0.025 mg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction