missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H5N1 Hemagglutinin Antibody (2B7), Biotin - (A/Vietnam/HN31242/2007), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22605B
This item is not returnable.
View return policy
Description
Influenza A H5N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H5N1 Hemagglutinin in Influenza A Virus H5N1 samples. It is validated for Western Blot, ELISA.
Specifications
| Influenza A H5N1 Hemagglutinin | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| HA | |
| Recombinant Influenza A H5N1 Hemagglutinin (17-338aa) purified from Baculovirus | |
| 0.1 mL | |
| Primary | |
| Anti-human H5N1/HA1 mAb, clone 2B7, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human H5N1/HA1 protein. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 2B7 | |
| Western Blot, ELISA | |
| HA, Hemagglutinin | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3654620 | |
| Influenza virus, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction