missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H5N1 HA Chimeric Recombinant Rabbit Monoclonal Antibody (310-18A3)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA551687

Product Code. 30088435

  • £477.00 / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific for the H5N1 and H1N1 subtypes of influenza A virus. Influenza viruses are negative-sense, single-stranded, segmented RNA viruses.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Influenza A H5N1 HA Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Rabbit
Protein A
RUO
Virus
Antibody
IgG κ
ELISA, Functional Assay
310-18A3
Unconjugated
flu hemagglutinin; influenza hemagglutinin
This antibody was raised by cloning antibody sequences form B cells isolated from people immunized with the H5 DNA vaccine.
200 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.