Learn More
Invitrogen™ Influenza A H3N2 Monoclonal Antibody (30-2F11-F7-A5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA183743
Description
MA1-83743 is specific for influenza A H3 antigen. This antibody may not react with all H3N2 Influenza A strains: it reacts strongly with the A/Shainghai/11/87 strain, but shows no reactivity with the A/Wuhan/359/95 strain. Mouse anti Influenza A H3 antigen antibody, clone 30-2F11-F7-A5recognizes the Influenza A H3 antigen and shows no reactivity to influenza B strains.
Influenza viruses consists of three types (Influenza A, B, and C), which make up three of the five genera in the Orthomyxoviridae family. Commonly know as the flu, influenza strains can affect both birds and mammals. Influenza strains form roughly spherical particles and are similar in composition with viral envelope containing two main types of glycoproteins (hemagglutinin and neuraminidase), wrapped around the central core of the viral RNA genome and other viral proteins that package and protect the RNA. Unusually for a virus, the influenzavirus genome is not a single piece of nucleic acid; rather a seven to eight segmented fragments of negative-sense RNA with each fragment expressing different viral proteins. Influenza spreads seasonally, resulting in about three to five million yearly cases of severe illness and about 250,000 to 500,000 deaths annually or into the millions in pandemic years.
Specifications
| Influenza A H3N2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| Immunocytochemistry | |
| 30-2F11-F7-A5 | |
| Unconjugated | |
| H3N2; Influenza H3N2 | |
| Influenza blend. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.