missing translation for 'onlineSavingsMsg'
Learn More

Influenza A H3N2 Hemagglutinin Antibody (1B7), Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18424486 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424486 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424486 Supplier Novus Biologicals Supplier No. NBP222553AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Influenza A H3N2 Hemagglutinin Monoclonal antibody specifically detects Influenza A H3N2 Hemagglutinin in Influenza A virus H3N2 samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H3N2 Hemagglutinin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1B7
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Recombinant Influenza A Virus Hemagglutinin (17-345aa) purified from Baculovirus
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Influenza A virus H3N2
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.